Nalgene™ Super Versi-Dry™ Surface Protectors
Nalgene™ Super Versi-Dry™ Surface Protectors
Actual product may vary
Thermo Fisher Scientific
Nalgene™ Super Versi-Dry™ Surface Protectors
Absorb spills and cushion breakable items with long-lasting, waterproof, skid-resistant and chemical-resistant surface protectors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensionsLxWDescription
74050-00Roll
74000-00250 ft. Dispenser Pk. Roll
74018-00Mat
74200-00Wide Roll
74218-00Mat
Catalog number 74050-00
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Roll
Absorb spills and cushion breakable items with Thermo Scientific™ Nalgene™ Super Versi-Dry™ Surface Protectors. They are long-lasting, waterproof, skid-resistant and chemical-resistant. The cloth-like top layer will not fall apart even when wet.
  • Cushioned surface minimizes breakage of glass labware when dropped or tipped over
  • Sheets lie flat and are skid resistant
  • Low-pile flat surface provides stability for glassware that is likely to tip
  • Polyethylene backing resists harsh chemicals and protects benchtop from spills and corrosion
  • Super Versi-Dry quickly absorbs up to 1050mL/m2 to quickly absorb spills with thousands of Thermo Scientific Thirsty Cells
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Product TypeSurface Protector
Absorbancy1050 mL/m2
Length (English)100 ft.
Length (Metric)30 m
Width (English)20 in.
Width (Metric)51 cm
DescriptionRoll
No. per Case4 Rolls
No. per Pack1 Roll
Item DescriptionVersi-Dry Super Absorbant Roll 20 in. x 100 ft.; 4 rolls x 100 ft.
Unit Size400