Nalgene™ Large Polypropylene Sterilizing Pans
Nalgene™ Large Polypropylene Sterilizing Pans
Actual product may vary
Nalgene™ Large Polypropylene Sterilizing Pans
Dry and sterilize instruments and labware with these large, polypropylene sterilizing pans, featuring rolled edges for extra stiffness and molded-in handles for easy carrying.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (L x W x H)
6900-002020 L21.375 x 17.125 x 5.125 in. (543 x 435 x 130 mm)
Catalog number 6900-0020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
20 L
Dimensions (L x W x H):
21.375 x 17.125 x 5.125 in. (543 x 435 x 130 mm)
Dry and sterilize instruments and labware with Thermo Scientific™ Nalgene™ Large Polypropylene Sterilizing Pans. These autoclavable pans are also useful for collecting and washing soiled lab equipment. Pans feature rolled edges for extra stiffness and molded-in handles for easy carrying.
  • Pans are tapered for nesting to save space
  • Heavy wall construction will withstand rugged use and repeated autoclaving
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Capacity (Metric)20 L
Dimensions (L x W x H)21.375 x 17.125 x 5.125 in. (543 x 435 x 130 mm)
DescriptionSterilizing pans are useful for drying and sterilizing instruments, labware; collecting and washing soiled lab equipment
Height (Metric)130 mm
Length (Metric)543 mm
MaterialPolypropylene
TypeSterilizing Pan
Width (Metric)435 mm
Length (English)21.375 in.
No. per Pack1
Height (English)5.125 in.
No. per Case6
ShapeRectangular
Width (English)17.125 in.
AutoclavableAutoclavable
Unit Size6