Nalgene™ Black Ink Lab Markers
Nalgene™ Black Ink Lab Markers
Actual product may vary
Nalgene™ Black Ink Lab Markers
Nalgene™ Black Ink Lab Markers
Nalgene™ Black Ink Lab Markers
Nalgene™ Black Ink Lab Markers
Thermo Fisher Scientific
Nalgene™ Black Ink Lab Markers
Mark on glass, plastic and metal with these black ink lab markers that have caps with a convenient packet clip.
Have Questions?
Catalog NumberDescriptionColor
6310-0010Lab Pen, Fine LineBlack
Catalog number 6310-0010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Lab Pen, Fine Line
Color:
Black
Mark on glass, plastic and metal with Thermo Scientific™ Nalgene™ Black Ink Lab Markers, recommended for use with Nalgene™ Polyolefin paper, notebooks and labels. These markers have caps with a convenient packet clip. Choose from fine point or broad line options.
  • Felt-tip marking pens with pocket clips and black ink
  • Choice between fine-line pen or broad-line marker
  • Writes on glass, plastic or metal
Specifications
Tip StyleFine
ColorBlack
DescriptionLab Pen, Fine Line
No. per Case12
No. per Pack4
Unit Size12