Globe Scientific 50mL Polypropylene Centrifuge Tube
Globe Scientific 50mL Polypropylene Centrifuge Tube
Actual product may vary
Thermo Fisher Scientific
Globe Scientific 50mL Polypropylene Centrifuge Tube
For centrifugation and storage of critical laboratory reagents including culture media, buffers and antibodies
Have Questions?
Catalog NumberCapacity (Metric)
624050 mL
Catalog number 6240
Price (MYR)
33,339.00
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
33,339.00
Add to cart
  • Feature flat-top, plug-seal leak resistant, red screw caps
  • Printed with white graduation marks in 0.5mL increments
  • White writing area for sample identification
  • Spin Rate: 10,000 RCF (12,320 x g)
  • Dimensions: 29.5 x 114mm
Specifications
Closure ColorRed
Diameter (English) Outside1.16 in.
Capacity (Metric)50 mL
MaterialPolypropylene
SterilityNon-sterile
Capacity (English)1.6 oz.
Closure IncludedYes
Diameter (Metric)29.5 mm
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF10,000 x g
Unit Size