Nalgene™ Versi-Dry™ Surface Protectors
Nalgene™ Versi-Dry™ Surface Protectors
Actual product may vary
Thermo Fisher Scientific
Nalgene™ Versi-Dry™ Surface Protectors
Quickly absorb spills and cushion breakable labware with these surface protectors. Use as a bench protector, shelf liner or cabinet liner to protect against the harshest of chemicals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Description
62060-00300 ft.Roll
62065-00
62080-00
Catalog number 62060-00
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
300 ft.
Description:
Roll
Quickly absorb spills and cushion breakable labware with Thermo Scientific™ Nalgene™ Versi-Dry™ Surface Protectors. Each surface protector is composed of thousands of Thermo Scientific™ Thirsty Cells™ for heightened performance. The polyethylene backing protects the benchtop from the harshest of chemicals. Use as a bench protector, shelf liner, cabinet liner or for wiping up isolated spills.
  • Cushioned surface minimizes breakage of glass labware when dropped or tipped over
  • Skid resistant polyethylene backing resists harsh chemicals and protects your benchtop from spills and corrosion
  • Low-pile flat surface provides stability for glassware that is likely to tip
  • Versi-Dry quickly absorbs up to 750mL/m2 spills with its thousands of Thirsty Cells
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Product TypeSurface Protector
MaterialPolyethylene
Absorbancy750 mL/m2
Length (English)300 ft.
Length (Metric)91 m
Width (English)20 in.
Width (Metric)51 cm
DescriptionRoll
No. per Case2 rolls
No. per Pack1 roll
Quantity2 Pk.
Item DescriptionVersi-Dry Standard Absorbancy Roll 20 in. x 300 ft.; 2 rolls x 300 ft.
Unit Size600