Histone H2BK15ac Polyclonal Antibody, Active Motif™
Histone H2BK15ac Polyclonal Antibody, Active Motif™
Actual product may vary
Histone H2BK15ac Polyclonal Antibody, Active Motif™
Histone H2BK15ac Polyclonal Antibody, Active Motif™
Histone H2BK15ac Polyclonal Antibody, Active Motif™
Rabbit Polyclonal Antibody
Have Questions?
Catalog NumberQuantity
61321100 μL
Catalog number 61321
Price (MYR)
2,546.00
Add to cart
Quantity:
100 μL
Price (MYR)
2,546.00
Add to cart
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
Specifications
AntigenHistone H2BK15ac
ApplicationsDot Blot, Western Blot
ClassificationPolyclonal
ConcentrationLot-Specific
ConjugateUnconjugated
FormulationPBS with 30% glycerol and 0.035% sodium azide; pH 7.5
GeneH2BC12
Gene Accession No.O60814, P23527, P62807, Q16778, Q5QNW6, Q6DN03, Q6DRA6, Q99877
Gene Alias2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K15; Acetyl-Histone H2B Lys15; AV127319; dJ193B12.2; dJ221C16.3; dJ221C16.8; GL105; H2B; H2b 613; H2B GL105; H2B histone family, member A; H2B histone family, member D; H2B histone family, member K; H2B histone family, member L; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member S; H2B histone family, member T; H2B K; H2B.1; H2B.1A; H2B.2; H2B/a; H2B/d; H2B/g; H2B/h; H2B/I; H2B/k; H2B/l; H2B/n; H2B/o; H2B/q; H2B/s; H2B/t; H2b-613; H2BC10; H2BC4; H2BC6; H2BC7; H2bc8; H2BFA; H2BFAiii; H2BFD; H2BFG; H2BFH; H2BFK; H2BFL; H2BFN; H2BFO; H2BFQ; H2bfs; H2BFT; H2BGL105; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2BLys15ac; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRIP1; Hist1h2ba; Hist1h2bc; Hist1h2be; HIST1H2BF; HIST1H2BG; HIST1H2BI; Hist1h2bk; HIST1H2BN; HIST1H2BO; HIST1H3I; HIST2H2BC; HIST2H2BD; HIST2H2BE; HIST2H2BF; histone 1, H2ba; histone 1, H2bc; histone 1, H2bg; histone 1, H2bi; histone 1, H2bk; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone cluster 1, H2ba; histone cluster 1, H2bc; histone cluster 1, H2bg; histone cluster 1, H2bi; histone cluster 1, H2bk; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2bc (pseudogene); histone cluster 2, H2bd (pseudogene); histone cluster 2, H2be; histone cluster 2, H2bf; histone family member; histone H2B; Histone H2B type 1-A; Histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; Histone H2B type 1-K; Histone H2B type 1-N; Histone H2B type 1-O; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B, testis; Histone H2B.1 A; Histone H2B.2; Histone H2B.a; Histone H2B.d; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.k; Histone H2B.l; Histone H2B.n; Histone H2B.q; Histone H2B-GL105; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; T25626; testis-specific histone 2b; Testis-specific histone H2B; Th2b
Gene SymbolsH2BC10, H2BC12, H2BC15, H2BC17, H2BC18, H2BC19P, H2BC20P, H2BC21, H2BC4, H2BC8
Host SpeciesRabbit
ImmunogenThis antibody was raised against a peptide including acetyl-lysine 15 of human histone H2B.
Purification MethodAntigen affinity chromatography
Quantity100 μL
Regulatory StatusRUO
Primary or SecondaryPrimary
Gene ID (Entrez)337873, 337874, 440689, 8339, 8341, 8346, 8347, 8348, 8349, 85236
Target SpeciesHuman
Product TypeAntibody
FormLiquid
IsotypeIgG
Unit Size
Contents & Storage
-20°C, Avoid Freeze/Thaw Cycles