Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Actual product may vary
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge, 36cc
Quickly attain and hold vacuum of 25 in. (635 mm) Hg with these hand-operated vacuum pumps featuring a new, more durable and ergonomic handle design.
Have Questions?
Catalog number 6131-0020
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quickly attain and hold vacuum of 25 in. (635 mm) Hg with Thermo Scientific™ Nalgene™ Repairable Hand-Operated PVC Body Vacuum Pumps without Gauge. These pumps feature a new, more durable and ergonomic handle design. This repairable economy version is ideal for field or lab use.
Easy and Convenient Operation
  • Polyvinyl Chloride (PVC) body
  • Ergonomic vacuum trigger release requires only one hand to operate and releases vacuum with the touch of an index finger
  • Adjustable vacuum release rate provides control
  • Full blank port pressure is obtained with only two strokes, making operation efficient
  • Removable cover on exhaust port and trigger to release vacuum without disconnecting pumps from the line
  • Economical design without vacuum gauge 
  • Adjustable vacuum release rate controlled by trigger on pump
  • Nozzle fits standard 0.25″ I.D. tubing
  • Can be disassembled for repair and replacement of internal parts (see catalog numbers for repair kits in the table below)
Includes:
Supplied with 2 ft. length of Clear Plastic Tubing
Specifications
DescriptionRepairable Hand Vacuum Pump
No. per Pack1
No. per Case4
Max. Vacuum25 in. Hg
Flow Rate36 cc/stroke
TypeVacuum Pump
For Use With6132-3000 Repair Kit
Unit Size4