Nalgene™ Acrylic Parafilm™ Dispenser
Nalgene™ Acrylic Parafilm™ Dispenser
Actual product may vary
Nalgene™ Acrylic Parafilm™ Dispenser
Nalgene™ Acrylic Parafilm™ Dispenser
Nalgene™ Acrylic Parafilm™ Dispenser
Nalgene™ Acrylic Parafilm™ Dispenser
Securely hold rolls for easy dispensing with the Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser.
Have Questions?
Catalog NumberDescription
5833-0001Acrylic Parafilm Dispenser
Catalog number 5833-0001
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Acrylic Parafilm Dispenser
Securely hold rolls for easy dispensing with the Thermo Scientific™ Nalgene™ Acrylic Parafilm™ Dispenser. This acrylic dispenser can hold one box of 4″ film or two boxes of 2″ film. A channeled front allows for cutting Parafilm.
Ordering Information:
Storage for blade is included but blade is not included.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Dimensions (L x W x H)6 x 4.75 x 6 in.
Length (Metric)152 mm
Width (Metric)121 mm
Height (Metric)152 mm
DescriptionAcrylic Parafilm Dispenser
Quantity1 Ea.
Unit SizeEACH