Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Actual product may vary
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Thermo Fisher Scientific
Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Carry out daily laboratory work with Thermo Scientific™ Heratherm™ General Protocol Ovens, featuring temperature uniformity, easy settings and low energy consumption.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)TypeElectrical Requirements
51029328419 LGravity Convection230 V 50/60 Hz
5102813965 LGravity Convection230 V 50/60 Hz
51028140105 LGravity Convection230 V 50/60 Hz
51028141176 LGravity Convection230 V 50/60 Hz
5102814866 LMechanical Convection Oven230 V 50/60 Hz
51028149104 LMechanical Convection Oven230 V 50/60 Hz
51028150179 LMechanical Convection Oven230 V 50/60 Hz
51029340774 LGravity Convection230 V 50/60 Hz
Catalog number 51029328
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
419 L
Type:
Gravity Convection
Electrical Requirements:
230 V 50/60 Hz
Carry out daily laboratory work with Thermo Scientific™ Heratherm™ General Protocol Ovens, featuring temperature uniformity, easy settings and low energy consumption. Designed for low energy consumption with unique door design and highly efficient, insulation-reducing heat emission. Additional features include a built in timer, easy-to-view display and an alarm system to protect samples.
Features:
  • Operating temperatures from 50° to 250°C
  • Automatic over-temperature alarm system
  • Low overall energy consumption
  • Built in timer
  • Corrosion-resistant stainless-steel chambers with rounded corners
  • Large, easy to read vacuum fluorescent display
  • Easy to use, microprocessor-controlled touch button operation
  • Doors can be opened over 180°C
  • Choice of gravity or mechanical convection models
Availability may vary by country.
Specifications
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)419 L
Temperature (Metric)50°C to 250°C
TypeGravity Convection
No. of Shelves2 supplied/39 max.
Temperature Stability±0.5°C
TimerON / OFF
Dimensions (D x W x H) Interior23.2 x 21.4 x 51.5 in. (59 x 54.4 x 130.7 cm)
Dimensions (L x W x H)30.3 x 30.6 x 60.8 in. (77 x 77.8 x 154.5 cm)
Length (English)30.3 in.
Length (Metric)77 cm
Width (English)30.6 in.
Width (English) Interior21.4 in.
Width (Metric)77.8 cm
Width (Metric) Interior54.4 cm
Height (English)60.8 in.
Height (English) Interior51.5 in.
Height (Metric)154.5 cm
Height (Metric) Interior130.7 cm
Depth (English) Interior23.2 in.
Depth (Metric) Interior59 cm
Electrical Requirements230 V 50/60 Hz
Voltage230 V
Wattage2400 w
Amperage10.5 A