Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Actual product may vary
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Heratherm™ Advanced Protocol Microbiological Incubators
Thermo Fisher Scientific
Heratherm™ Advanced Protocol Microbiological Incubators
Perform drying applications with our advanced protocol microbiological incubators, which include dual-convection technology and delivers excellent temperature consistency.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)VoltageTemperature Range (Metric)Temperature Stability
5102813366 L230 VAmbient +5° to 105°C±0.1°C
5102871766 L230 VAmbient +5° to 105°C±0.1°C
51028134104 L230 VAmbient +5° to 105°C±0.1°C
51028718104 L230 VAmbient +5° to 105°C±0.1°C
51028135178 L230 VAmbient +5° to 105°C±0.1°C
Catalog number 51028133
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
66 L
Voltage:
230 V
Temperature Range (Metric):
Ambient +5° to 105°C
Temperature Stability:
±0.1°C
Perform drying applications with Thermo Scientific™ Heratherm™ Advanced Protocol Microbiological Incubators, which include dual-convection technology and deliver excellent temperature consistency. These incubators are an effective and flexible tool in the lab and can be used for multiple applications.
Features:
  • Dual convection for application versatility — fan speed adjustable from 0 to 100%
  • Advanced digital timer for daily or weekly ON/OFF cycles
  • Internal socket (120V models only) allows the use of equipment, such as shakers or stirrers, inside the unit
  • Easy-to-clean, corrosion-resistant stainless-steel interior (AISI 304)
  • Intuitive user interface for setting temperature
  • Large, easy-to-read vacuum fluorescent display
  • Exceptionally small footprint
Temperature:
  • Broad temperature range from 5°C above ambient to 105°C — suitable for drying applications
  • Temperature uniformity of ±0.6 (fan off) and between ±0.2°C and ±0.4C (fan full speed) — measured at 37°C
  • Temperature stability of ±0.1°C — measured at 37°C
  • Internal glass door allows sample viewing without impacting temperature
Ordering Information:
Products only available in North America
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)2.3 cu. ft.
Capacity (English) Shelf55 lb.
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)66 L
Convection TechnologyDual convection
No. of Shelves2 supplied/ 13 max.
Plug TypePlug type provided based on country
Temperature Range (Metric)Ambient +5° to 105°C
TypeMicrobiological Incubator
Temperature Stability±0.1°C
Temperature Uniformity±0.6°C
Timerweekly / real time / hour
Dimensions (D x W x H) Interior14.5 x 13.9 x 20 in. (36.8 x 35.4 x 50.8 cm)
Dimensions (L x W x H)22.2 x 20.9 x 28.3 in. (56.5 x 53 x 72 cm)
Length (English)22.2 in.
Length (Metric)56.5 cm
Width (English)20.9 in.
Width (English) Interior13.9 in.
Width (Metric)53 cm
Width (Metric) Interior35.4 cm
Height (English)28.3 in.
Height (English) Interior20 in.
Height (Metric)72 cm
Height (Metric) Interior50.8 cm
Depth (English) Interior14.5 in.
Depth (Metric) Interior36.8 cm
Weight (English)99 lb.
Weight (Metric)45 kg
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Wattage850 w
Amperage3.7 A