Heratherm™ General Protocol Microbiological Incubators
Heratherm™ General Protocol Microbiological Incubators
Actual product may vary
Heratherm™ General Protocol Microbiological Incubators
Heratherm™ General Protocol Microbiological Incubators
Thermo Fisher Scientific
Heratherm™ General Protocol Microbiological Incubators
Perform a range of general applications with these general protocol incubators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltageCapacity (Metric)
51028132230 V194 L
51029334230 V747 L
51028130230 V75 L
51028131230 V117 L
51029322230 V405 L
Catalog number 51028132
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Voltage:
230 V
Capacity (Metric):
194 L
Use Thermo Scientific™ Heratherm General Protocol Microbiological Incubators for routine applications. The Heratherm General Protocol Incubator line is designed with sample safety in mind, for applications in pharmaceutical, medical, food and research laboratories.
  • Gravity convection provides gentle air flow and minimal drying out
  • Temperature range from ambient 5° to 75°C
  • Temperature uniformity of ±0.6°C (measured at 37°C)
  • Temperature stability of ±0.2°C (measured at 37 °C)
  • Corrosion-resistant stainless-steel chamber (AISI 430)
  • Intuitive user interface for setting temperature
  • Large, easy to read vacuum fluorescent display
  • Internal glass door allows sample viewing without impacting temperature
  • Exceptionally small footprint
Ordering Information:
Availability may vary by country.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)6.9 cu. ft.
Certifications/ComplianceCE Marked
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)194 L
Chamber MaterialStainless Steel
Convection TechnologyGravity convection
No. of Shelves2 supplied/ 19 max.
Plug TypeCountry-specific
Temperature Range (Metric)Ambient +5° to 75°C
TypeMicrobiological Incubator
Dimensions (D x W x H) Interior23.2 x 18.3 x 27.9 in. (58.9 x 46.4 x 70.8 cm)
Dimensions (L x W x H)29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
Weight (English)143 lb.
Weight (Metric)65 kg
Electrical Requirements230 V, 50/60 Hz
Voltage230 V
Wattage710 w
Amperage3.1 A