Sterilin™ 30 to 140mm Petri Dishes
Sterilin™ 30 to 140mm Petri Dishes
Actual product may vary
Sterilin™ 30 to 140mm Petri Dishes
Sterilin™ 30 to 140mm Petri Dishes
Sterilin™ 30 to 140mm Petri Dishes
Thermo Fisher Scientific
Sterilin™ 30 to 140mm Petri Dishes
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140mm Petri Dishes. These small dishes are suitable for most applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)Vent
501V139 mm21.1 mm3
FKS11339283139 mm21.1 mm3
Catalog number 501V
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
139 mm
Height (Metric):
21.1 mm
Vent:
3
Save incubator space with Thermo Scientific™ Sterilin™ 30 to 140mm Petri Dishes. These small dishes are suitable for most applications.
NOT AVAILABLE IN AMERICA
  • 50mm diameter dish (catalog number 124) is over 20mm deep and designed for use with liquid media to aid handling
  • 55mm dish accommodates 47mm membrane filters making it suitable for water testing
  • 140mm dish is produced and tested in accordance with the EN ISO 24998 Petri dish standard which includes stringent dimensional controls
Specifications
ShapeRound
SterilityNon-sterile
Surface TreatmentNon-treated
TypePetri Dish
No. per Case80
No. per Pack10
Vent3
Height (Metric)21.1 mm
Diameter (Metric)139 mm
Unit Size80