Water Purification Systems Storage Reservoirs
Water Purification Systems Storage Reservoirs
Actual product may vary
Thermo Fisher Scientific
Water Purification Systems Storage Reservoirs
Safely store purified water from your Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ system with one of these compatible storage reservoirs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Description
5015550060 LStorage Reservoir, with level display
5015549530 LStorage Reservoir, with level display
06.500130 to 100 LSterile Overflow for Storage Reservoir Tank
5015549960 LStorage Reservoir, with level display and recirculation pump
5015549430 LStorage Reservoir, with level display and recirculation pump
5015550160 LStorage Reservoir, 60L
5015549730 LStorage Reservoir
Catalog number 50155500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
60 L
Description:
Storage Reservoir, with level display
Safely store purified water from a compatible system with Thermo Scientific™ Water Purification Systems Storage Reservoirs. For use with Thermo Scientific™ Barnstead™ GenPure™, Pacific™ or Smart2Pure™ systems, these reservoirs are compatible with the following models: GenPure, GenPure Pro, GenPure xCAD, GenPure xCAD Plus, Pacific RO, Pacific TII and Smart2Pure.
Features:
  • Large opening offers easy and effective cleaning.
  • Optional safety filters include a CO2 absorber prevents conductivity increases from drawn-in CO2 and a sterile overflow to prevent microorganisms from entering and contaminating reservoir water.
  • Polyethylene reservoir is constructed with inert materials opaque to light.
  • Recirculation pump (optional) protects water from bacterial growth during standstill while maintaining a low conductivity value.
  • Wall mount is optional.
  • Conical bottom outlet allows for complete draining.
For use with GenPure, GenPure Pro, and GenPure xCAD purification systems only:
  • 06.5038
  • 06.5068
For use with Pacific RO water purification systems only:
  • 06.5034
  • 06.5064
  • 06.5084
For use with Pacific TII water purification systems only:
  • 06.5032
  • 06.5062
  • 06.5082
For use with Pacific RO & Pacific TII water purification systems only:
  • 06.5033
  • 06.5063
  • 06.5083
For use with Smart2Pure 12 water purification systems only:
  • 06.5040
  • 06.5070
Specifications
Capacity (Metric)60 L
DescriptionStorage Reservoir, with level display
For Use With (Equipment)Pacific RO, Pacific TII
Unit Size