Ion Exchange Stainless Steel (pressure resistant) Cartridges
Ion Exchange Stainless Steel (pressure resistant) Cartridges
Actual product may vary
Thermo Fisher Scientific
Ion Exchange Stainless Steel (pressure resistant) Cartridges
Ready-to-connect for 300 to 3000L/hr requirements, these stainless-steel cartridges are practice-proven mixed bed systems for fully demineralized water.
Have Questions?
Catalog NumberDescriptionFlow RateCapacityDiameter (Metric)
50155444DI 2000 Ion Exchange Stainless Steel (pressure resistant) Cartridges300 L/hr.2000 L240 mm
Catalog number 50155444
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
DI 2000 Ion Exchange Stainless Steel (pressure resistant) Cartridges
Flow Rate:
300 L/hr.
Capacity:
2000 L
Diameter (Metric):
240 mm
Ready-to-connect for 300 to 3000L/hr requirements, Thermo Scientific™ Ion Exchange Stainless Steel Cartridges are practice-proven mixed bed systems for fully demineralized water.
Optimal performance is ensured by the flow design and the quality-controlled ion exchange resins. Provide fully demineralized water with only a very low conductivity.
To function reliably, ion exchange cartridges require not only high quality mixed bed resins, but also a reliable regeneration technology coupled with high-grade chemicals. Our regeneration services feature recorded evidence of the capacity and quality for every batch created. These records include:
  • Receiving inspection
  • Batch test record
  • TOC-Monitoring
  • DIN EN ISO 9001 : 2000
Common Specifications for all deionizers include:
  • All cartridges are pressure resistant up to 10 bar
  • Water temperature maximum is 30°C
  • Material for the cartridge is stainless steel V4A
  • Connectors are R3/4 in.
  • Wall mount for each cartridge is not included, but can be ordered using part number 03.1405
Specifications
DescriptionDI 2000 Ion Exchange Stainless Steel (pressure resistant) Cartridges
Capacity2000 L
Flow Rate300 L/hr.
MaterialStainless Steel
Purification MethodIon Exchange
Resin TypeMixed Bed
Purity0.1 to 20 μS/cm
TypeIon Exchange Cartridge
Height (Metric)490 mm
Diameter (Metric)240 mm
Net Weight (Metric)18 kg
Unit Size