Storage Canes
Storage Canes
Actual product may vary
Storage Canes
Storage Canes
Storage Canes
Thermo Fisher Scientific
Storage Canes
Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDesignLength (Metric)
5015-0001Cryo Storage CaneStepped30 cm
378441Cryo Storage CaneStandard29 cm
FKS10100341Cryo Storage CaneStepped30 cm
5015-0002Cryo Storage CaneStandard30 cm
Catalog number 5015-0001
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Cryo Storage Cane
Design:
Stepped
Length (Metric):
30 cm
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
  • Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
  • Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
Specifications
DesignStepped
MaterialAluminum
TypeCryo Storage Cane
Quantity12/Pk., 48/Cs.
Length (English)11.31 in.
Length (Metric)30 cm
Unit Size48