Barnstead™ Point of Use Water Purification Systems Ultrafilters
Barnstead™ Point of Use Water Purification Systems Ultrafilters
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Point of Use Water Purification Systems Ultrafilters
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
50133980GenPure, GenPure Pro, GenPure xCAD
50133981MicroPure, Smart2Pure (UF and UF/UV units only)
Catalog number 50133980
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
GenPure, GenPure Pro, GenPure xCAD
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
Specifications
DescriptionUltrafilter; For use with GenPure, GenPure Pro, GenPure xCAD (UF, UF/UV and UF/UV-TOC units only)
For Use With (Equipment)GenPure, GenPure Pro, GenPure xCAD
TypeUltrafilter
Unit Size