Barnstead™ Pacific™ RO Water Purification System
Barnstead™ Pacific™ RO Water Purification System
Actual product may vary
Thermo Fisher Scientific
Barnstead™ Pacific™ RO Water Purification System
Improve functional and economical production of high-purity water with this modularly designed laboratory reverse osmosis system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPermeate performance
501323867 L/hr.
5013238712 L/hr.
5013238820 L/hr.
Catalog number 50132386
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Permeate performance:
7 L/hr.
Improve functional and economical production of high-purity water with the Thermo Scientific™ Barnstead™ Pacific™ RO Water Purification System. This modularly designed laboratory reverse osmosis system was specifically developed for functional and economical production of high-purity water.
Product Features
  • Five system options with permeate flows of 3, 7, 12, or 20, or 40L/hr
  • Space-saving design allows system to sit on the laboratory bench or be mounted on the wall
  • Simple system upgrade accommodates future pure water capacity requirements
Reliable Operation
  • Microprocessor control offers automatic operation with continual monitoring of all important parameters
  • Required filling level of the reservoir can be customized
  • Automatic return to operating mode ensures recirculation during periods of standstill
Easy-to-operate Clear Display
  • Resistivity/conductivity clearly displayed on large back-lit control panel that tilts for optimum viewing
  • Status of current operating mode clearly indicates 'production', 'stand-by' and 'cleaning' modes
  • Reservoir fill-level shown as %
Integrated Feed Water Monitoring
Good quality feed water is a prerequisite for high purity water and extends cartridge service life
GLP-compliant Documentation
Real-time clock records system errors and faults
Ongoing data storage collects four weeks information displayed in clear text
Operating system is code-protected to prevent unauthorized changes to system settings
RS-232 interface with adjustable sender interval allows for customized timing of data transmission of measured values and faults to a PC or data printer
Cell constant of 0.16 cm-1 ensures precise conductivity measurements
Pacific RO Pretreatment
Two 10 in. (25.4cm) cartridge filter housings with a 5Glc mum prefilter plus activated carbon to prevent entry of particles >5Glc mum and high concentrations of free chlorine — includes hardness stabilizer cartridge which binds calcium and prevents its precipitation.
Worry-free Pure Water Storage
Reservoir are constructed of pigment-free polyethylene
Conical bottom allows for easy cleaning, disinfecting and complete emptying
Optional Hand Dispenser
Gain increased flexibility by adding a hand dispenser with 3 meter cord which connects to tank. Use only with a tank equipped with a pressure pump (06.5032, 06.5062, 06.5082).  Ships with a 0.2 micron final filter. (Part #50138221)
Warranty: 1 year, parts and labor
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Bacterial retention99%
DescriptionBarnstead Pacific RO 7
Feed Water MonitoringIntegrated
Flow Rate7 L/hr.
TypeReverse Osmosis Water Purification System
Feedwater Connector3/4 in. NPT
Operating Pressure1.5 to 6 bar
Permeate performance7 L/hr.
Power Consumption0.1 kW
Temperature Range (Metric)2°C to 35°C
Width (Metric)372 mm
Height (Metric)603 mm
Depth (Metric)330 mm
Net Weight (Metric)21 kg
Electrical Requirements230 V 50/60 Hz