Shelves for Heratherm™ Ovens and Incubators
Shelves for Heratherm™ Ovens and Incubators
Actual product may vary
Thermo Fisher Scientific
Shelves for Heratherm™ Ovens and Incubators
Perforated and wire mesh shelves
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (English)Width (English)For Use With (Equipment)Length (English)
5012777412.9 in.12.9 in.Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 100l17.2 in.
5012777212.9 in.12.9 in.Heratherm General Protocol Microbiological Incubator 180l12.9 in.
5012777014.8 in.14.8 in.Heratherm General Protocol Microbiological Incubator 60l12.9 in.
5012776319.8 in.19.8 in.Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l17.2 in.
5012777719.8 in.19.8 in.Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l17.2 in.
5013524121.06 in.21.06 in.Heratherm Advanced Protocol Security Incubators 400l imh400-s and General Protocol Incubator 400l20.7 in.
5013524221.06 in.38.74 in.Heratherm Advanced Protocol Security Incubators 750l imh750-s and General Protocol Incubator 750l
5013524521.06 in.21.06 in.Heratherm Advanced Protocol Oven 400I OMH 400 and General Protocol Incubator 400l IGS400
5013524621.06 in.21.06 in.Heratherm Advanced Protocol Oven 750l OMH 750 and General Protocol Incubator 750l IGS750
Catalog number 50127774
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Depth (English):
12.9 in.
Width (English):
12.9 in.
For Use With (Equipment):
Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 100l
Length (English):
17.2 in.
Perforated and wire mesh shelves
Specifications
For Use With (Equipment)Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 100l
TypeShelf
Includes(2) Shelf Supports
MaterialStainless Steel
No. of Shelves2
Length (English)17.2 in.
Width (English)12.9 in.
Width (Metric)32.7 cm
Depth (English)12.9 in.
Depth (Metric)32.7 cm
Unit Size