Check Order Status
Stainless Steel Tubing, Outer Diameter: 1/16 in.
Actual product may vary
Thermo Fisher Scientific
Stainless Steel Tubing, Outer Diameter: 1/16 in.
Ideal for all plumbing in UHPLC systems
Have Questions?
Catalog Number
Grade
Diameter (English) Inner
Overall Length (Metric)
50127771
Premium
0.007 in.
3 m
Catalog number
50127771
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Grade:
Premium
Diameter (English) Inner:
0.007 in.
Overall Length (Metric):
3 m
Made from 316 Stainless Steel
Outer Diameter: 1/16 in.
Standard-grade tolerances: ±0.05 mm (.002 in.) O.D. & I.D.
Premium-grade tolerances: ±0.05 mm (.002 in.) O.D. & ±0.025 mm (.001 in.) I.D.
Specifications
Grade
Premium
Tolerance
±0.05 mm (0.002 in.) O.D. & ±0.025 mm (0.001 in.) I.D.
For Use With (Application)
Plumbing
Diameter (English) Inner
0.007 in.
Diameter (English) Outer
1/16 in.
Overall Length (Metric)
3 m
Material
Stainless Steel
Unit Size