Heratherm™ Compact Microbiological Incubators
Heratherm™ Compact Microbiological Incubators
Actual product may vary
Thermo Fisher Scientific
Heratherm™ Compact Microbiological Incubators
Save space with our compact microbiological incubator, which is ideal for a personalized workspace or crowded labs with small sample volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug Type
50125590Nema 5-15 US
50125882EU standard, UK and Australia cable/plug
Catalog number 50125590
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Plug Type:
Nema 5-15 US
Save space with the Thermo Scientific™ Heratherm™ Compact Microbiological Incubator. The extremely small footprint makes it ideal for a personalized workspace or crowded labs with small sample volumes. The most compact unit of the Heratherm microbiological incubators family has an 18L capacity, and operates in temperatures at or below ambient.
Features:
  • Minimal benchtop space for space restricted lab areas
  • Easy-to-use interface
  • Internal light and window in door facilitates sample observation
Temperature Security:
  • Temperatures at or below ambient: 17°C to 50°C
  • Temperature uniformity of ±1.2°C — measured at 37°C
  • Temperature stability of ± 0.2°C — measured at 37°C
  • High temperature accuracy
Ordering Information:
Availability may vary by country.
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)0.65 cu. ft.
Capacity (English) Shelf4.4 lb.
Certifications/CompliancecCSAus, CE
Controller TypeLED display
Capacity (Metric)18 L
Convection TechnologyMechanical convection
No. of Shelves2 supplied/ 3 max.
Plug TypeNema 5-15 US
Temperature Range (Metric)17°C to 50°C
TypeMicrobiological Incubator
Temperature Stability±0.2°C
Temperature Uniformity±1.2°C
Dimensions (D x W x H) Interior11.4 x 7.1 x 12.2 in. (29 x 18 x 31 cm)
Dimensions (L x W x H)18.5 x 10.2 x 16.3 in. (47 x 26 x 41.5 cm)
Length (English)18.5 in.
Length (Metric)47 cm
Width (English)10.2 in.
Width (English) Interior7.1 in.
Width (Metric)26 cm
Width (Metric) Interior18 cm
Height (English)16.3 in.
Height (English) Interior12.2 in.
Height (Metric)41.5 cm
Height (Metric) Interior31 cm
Depth (English) Interior11.4 in.
Depth (Metric) Interior29 cm
Weight (English)15.9 lb.
Weight (Metric)7.2 kg
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Wattage45 w
Amperage0.45 to 0.85 A