Heraguard™ ECO Clean Bench Floor Stands
Heraguard™ ECO Clean Bench Floor Stands
Actual product may vary
Thermo Fisher Scientific
Heraguard™ ECO Clean Bench Floor Stands
Choose from fixed, manual and electrically adjustable floor stands for the Thermo Scientific™ Heraguard™ ECO clean bench.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
50109312Adjustable Floor StandHeraguard ECO 1.2
50109311Floor StandHeraguard ECO 1.8, 1.8/95
50109977CastorsHeraguard ECO 0.9, 1.2, 1.5, 1.8
50109309Floor StandHeraguard ECO 1.2
50116442Floor StandHeraguard ECO 1.5
Catalog number 50109312
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Adjustable Floor Stand
For Use With (Equipment):
Heraguard ECO 1.2
Choose from fixed, manual and electrically adjustable Thermo Scientific™ Heraguard™ ECO Clean Bench Floors Stands for the Heraguard™ ECO clean bench.
† Due to DC motors, Heraguard™ ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
For Use With (Equipment)Heraguard ECO 1.2
DescriptionProvides variable ECO work height of 760 mm - 960 mm in 50 mm adjustable increments.
Product TypeAdjustable Floor Stand
Height Overall (Metric)680 to 880 mm
Unit Size