Cimarec i Poly 15 and Multipoint Stirrers
Cimarec i Poly 15 and Multipoint Stirrers
Actual product may vary
Thermo Fisher Scientific
Cimarec i Poly 15 and Multipoint Stirrers
Conduct stirring applications with these stirrers, which are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModel
50093557Multipoint 6
50093538Multipoint 15
50094596Poly 15
Catalog number 50093557
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Model:
Multipoint 6
Conduct stirring applications with Thermo Scientific™ Cimarec™ i Poly 15 and Multipoint Stirrers for reliable, safe performance. These stirrers are 100% maintenance- and wear-free, in a fully synchronized multipoint platform.
Features:
  • Large, flat, easy-to-clean work surfaces
  • Precise microprocessor control with bright digital display
  • Easily recall last used settings
  • Gradual start acceleration is gentle and ensures good magnetic coupling
  • Inductive drive ensures completely synchronized stirring at all positions
  • Operating conditions are -10°C to +40°C at 95%RH
Poly 15 Multipoint Stirrer:
  • Rugged, powder-coated, stainless-steel housing
  • Stir speeds up to 990rpm
  • Suitable for 15 × 250mL beakers or 6 × 1L flasks
  • IP32 protection rated
Advanced Multipoint 6/15 Stirrers:
  • Chemical-resistant, sealed stainless-steel housing
  • Adjustable power settings are separate from the speed control that decreases power consumption and lowers heat output
  • Smooth and even stirring as low as 80rpm
  • Stir speeds up to 2000rpm
  • Stir volumes up to 3L per multipoint
  • IP62 protection rated; easily cleaned with running water
Includes:
All models are 100-240V and contain a cord set with various plugs.
Recommended for:
Mass screening applications and other applications requiring multiple stirring points
Warranty: 5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed80 to 2000 rpm
Stirring Positions6
Top Plate MaterialStainless Steel
Capacity3 L
DisplayDigital
ShapeSquare
Voltage100/240 V
Wattage5/10/15/20 w
Dimensions (L x W x H)16.5 x 9.4 x 1.4 in. (420 x 240 x 35 mm)
Height (English)1.4 in.
Height (Metric)35 mm
Length (English)16.5 in.
Length (Metric)420 mm
Width (English)9.4 in.
Width (Metric)240 mm
Certifications/ComplianceCE
DescriptionMultipoint 6; Capacity 3L per multipoint; 6 stirring positions; 80 to 2000rpm
ModelMultipoint 6
Protection ClassIP62
Temperature Range (Metric)-10°C to +56°C (Operating)
TypeStirrer
Weight (English)16.5 lb.
Weight (Metric)7.5 kg
Unit Size