Finnpipette™ F1 Variable Volume Pipettes
Finnpipette™ F1 Variable Volume Pipettes
Actual product may vary
Thermo Fisher Scientific
Finnpipette™ F1 Variable Volume Pipettes
Maximize user comfort and reduce strain with the F1 system, which delivers performance with precision, all in a lightweight design. It's the ideal system for applications using expensive reagents and valuable samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Color Code
4641010N0.2 to 2 μLPink
4641020N0.5 to 5 μLPink
4641060N2 to 20 μLYellow
4641070N10 to 100 μLYellow
4641080N20 to 200 μLYellow
4641100N100 to 1000 μLBlue
4641110N0.5 to 5 mLGreen
4641120N1 to 10 mLRed
Catalog number 4641010N
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
0.2 to 2 μL
Color Code:
Pink
Enjoy the enhanced volume adjustment on Thermo Scientific™ Finnpipette™ F1 Variable Volume Pipettes, which is now silent and effortless with textured adjustment knob for a firmer feel when setting volumes. The adjustable finger rest and lightweight design maximize comfort without sacrificing performance
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifications
Compatible TipsFinntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
DescriptionFinnpipette F1
ColorPink
Number of Channels1
TypeVariable Volume Pipette
Volume (Metric)0.2 to 2 μL
Increments0.002μL
Color CodePink
Unit SizeEACH