Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Actual product may vary
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Thermo Fisher Scientific
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Support every day pipetting in comfort and with ideal function using these manual variable volume pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)dimColorCode
4640070
46400001 to 10 μL
46400101 to 10 μL
4640020
4640030
4640050
4640060
4640080
4640040
Catalog number 4640070
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Thermo Scientific™ Finnpipette™ F3 Variable Volume Single Channel Pipettes are comfortable to use, and provide ideal functionality in every day pipetting. The Finnpipette F3 pipette features a wide, color-coded supportive finger rest and ergonomic handle design. Finnpipette F3 pipettes are available in single and multichannel models.
Ergonomic Comfort
  • Wide color-coded supportive finger rest for identification support and ergonomics
  • Designed handle softly contours the shape of your hand
  • Large Ergovisio Display for volume recognition
Ideal Function
  • Durable handle material
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Advanced volume gearing mechanism for enhanced precision and accuracy
  • Super blow-out for efficient delivery of micro-size drops in 50ul models and below
Compatible with Finntip pipette tips, and other tip models
Specifications
Precision0.8 to 0.2%
Unit SizeEACH