Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Color | Compatible Tips |
---|---|---|---|
46200600 | 100 to 1000 μL | Blue | Finntip 1000, 1000 Ext., Flex 1000 |
46200400 | 10 to 100 μL | Yellow | Finntip Flex 200, 250 Univ, 200 Ext. |
46200700 | 0.5 to 5 mL | Green | Finntip 5 mL |
46200800 | 1 to 10 mL | Red | Finntip 10 mL, Flex 10 mL Ext. |
Features
Ergonomic Design
Compatible with:
Ideal for use with Thermo Scientific Finntip™ Flex or Finntip pipette tips.
Warranty:
2 years with web registration