Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Actual product may vary
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Injection Port Liners for GC Instruments
Use these injection port liners to optimize performance of Agilent GC instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Diameter (Metric) InnerDiameter (Metric) OuterPacking Material
453A1335Direct Straight Liner1.2 mm6.3 mmNone
453A1295Split Straight Liner4 mm6.3 mmNone
HYP453A1295Split Straight Liner4 mm6.3 mmNone
453A1315Split/Splitless FocusLiner with Single Taper4 mm6.3 mmQuartz Wool
453A1345Split/Splitless Liner with Single Taper4 mm6.3 mmNone
453A1925Splitless Liner with Single Taper4 mm6.3 mmQuartz Wool
453A2265Split Straight Liner4 mm6.3 mmQuartz Wool
HYP453A2265Split Straight Liner4 mm6.3 mmQuartz Wool
Catalog number 453A1335
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Application):
Direct Straight Liner
Diameter (Metric) Inner:
1.2 mm
Diameter (Metric) Outer:
6.3 mm
Packing Material:
None
Optimize the performance of Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
  • Highly deactivated borosilicate glass
  • Quartz wool packing material
  • IDxODxL: 4 x 6.3 x 78.5 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description453A1335
For Use With (Application)Direct Straight Liner
Quantity5
Length (Metric)78.5 mm
Packing MaterialNone
Product TypeDirect Straight Liner
Diameter (Metric) Inner1.2 mm
Diameter (Metric) Outer6.3 mm
For Use With (Equipment)GC Injection Port
Liner MaterialHighly Deactivated Borosilicate Glass
Unit Size