Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Actual product may vary
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Thermo Fisher Scientific
Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionCapacity (Metric)
4301-1000Separatory Funnel FEP 1000mL, ETFE Closure1000 mL
4301-2000Separatory Funnel FEP 2000mL, ETFE Closure2000 mL
FIS10-437-25DSeparatory Funnel FEP 1000mL; 53mm closure1000 mL
Catalog number 4301-1000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Separatory Funnel FEP 1000mL, ETFE Closure
Capacity (Metric):
1000 mL
Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.
Features:
  • Break-resistant and impervious to any chemical used in a separatory funnel
  • No phthalates to contaminate analyses
  • Useful in radiological and hazardous separations
  • Autoclavable/Leakproof† when used with provided Nalgene closure
  • Stem length: 65mm
Ordering Information:
For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.
Note:
Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.
Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)1000 mL
MaterialTeflon FEP
Height (Metric)371 mm
DescriptionSeparatory Funnel FEP 1000mL, ETFE Closure
Unit Size4