Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Actual product may vary
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Nalgene™ Polypropylene Analytical Funnels
Use standard filter papers with these funnels, featuring long stems and external ribbing that eliminates air lock.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopCapacity (Metric)
4250-006563 mm64 mL
4250-005554 mm41 mL
FKS1028454198 mm254 mL
Catalog number 4250-0065
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
63 mm
Capacity (Metric):
64 mL
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
  • Body of funnel is at a 60° angle with internal ribs sloped at 58° for rapid filtration
  • Autoclavable
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Diameter (Metric)Top63 mm
Capacity (Metric)64 mL
Diameter (Metric) Stem7 mm
MaterialPolypropylene
AutoclavableAutoclavable
DescriptionFunnel Analytical PP 65mm
Length (Metric) Stem58 mm
Quantity12/Pk.; 36/Cs.
Unit Size36