Benchtop Dewar Flasks
Benchtop Dewar Flasks
Actual product may vary
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Thermo Fisher Scientific
Benchtop Dewar Flasks
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
4150-20002 L
4150-900010 L
Catalog number 4150-2000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2 L
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.
  • High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
  • Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
  • Can be used from -196°C to +100°C
  • Pour spout and bottom grip area for ease of removal of used liquids
  • Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifications
No. per Case2
No. per Pack1
Capacity (English)67.628 fl oz.
Capacity (Metric)2 L
Depth (Metric) Interior231 mm
Depth (English) Interior9.1 in.
Diameter (Metric) Inner Mouth122mm
Diameter (English) Inner Mouth4.8 in.
Height (Metric)282mm
Height (English)11.1 in.
Unit Size2