Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Actual product may vary
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Thermo Fisher Scientific
Racks for Thermo Scientific™ 5 Shelf TSX/TDE Series Freezers
Maximize tube storage capacity per freezer with a racking solution for five inner-door, ultra-low temperature freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)HoldsNoofRacksPositions
39832617.3 and 23 cu. ft. capacity upright freezersBoxes (2 in.)16 Boxes/Rack
39833117.3 and 23 cu. ft. capacity upright freezersBoxes (2 in.)16 Boxes/Rack
FKS1136932117.3 and 23 cu. ft. capacity upright freezersBoxes (2 in.)16 Boxes/Rack
Catalog number 398326
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
17.3 and 23 cu. ft. capacity upright freezers
Holds:
Boxes (2 in.)
Positions:
16 Boxes/Rack
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
  • Warranty varies by country, please contact us for details
Specifications
Product TypeSide Access Freezer Rack (Adjustable)
Description2 in. Box Racks - Adjustable side access rack
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions16 Boxes/Rack
Shelves4 Tiers
For Use With (Equipment)17.3 and 23 cu. ft. capacity upright freezers
Width (Metric)13.7 cm
Width (English)5.4 in.
Height (Metric)23.9 cm
Height (English)9.4 in.
Depth (English)22.1 in.
Depth (Metric)56.1 cm
Unit Size