Chest Freezer Racks
Chest Freezer Racks
Actual product may vary
Thermo Fisher Scientific
Chest Freezer Racks
Use Thermo Scientific™ Chest Freezer Racks with Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Holds
3981842 in. Box Rack - Side Access Rack (Tower)3 cu. ft. chest freezersBoxes (2 in.)
189001Wire Basket Rail Set12.7cu. ft. freezersGeneral Freezer Items
3981862 in. Box Rack - Side Access Rack (Tower)13, 17, and 20 cu. ft. freezersBoxes (2 in.)
Catalog number 398184
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
2 in. Box Rack - Side Access Rack (Tower)
For Use With (Equipment):
3 cu. ft. chest freezers
Holds:
Boxes (2 in.)
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.
  • Warranty varies by country, please contact us for details
Specifications
Product TypeSide Access Rack Tower
Description2 in. Box Rack - Side Access Rack (Tower)
ColorSilver
HoldsBoxes (2 in.)
MaterialStainless Steel
Positions7 Boxes/Rack
For Use With (Equipment)3 cu. ft. chest freezers
Freezer Compatibility (Capacity)3 cu. ft.
Width (Metric)14 cm
Width (English)5.5 in.
Height (Metric)38.6 cm
Height (English)15.2 in.
Depth (English)5.5 in.
Depth (Metric)14 cm
Unit Size