Nalgene™ Polypropylene Graduated Cylinders
Nalgene™ Polypropylene Graduated Cylinders
Actual product may vary
Nalgene™ Polypropylene Graduated Cylinders
Nalgene™ Polypropylene Graduated Cylinders
Thermo Fisher Scientific
Nalgene™ Polypropylene Graduated Cylinders
Conduct applications accurately with these graduated cylinders, which feature generous pour spouts and molded-in graduations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
3662-10001000 mL10 mL
3662-0100100 mL1 mL
FIS08-572D100 mL1 mL
3662-0500500 mL5 mL
FIS08-572F500 mL5 mL
FIS08-572G1000 mL10 mL
FIS08-572H2000 mL20 mL
Catalog number 3662-1000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Graduations:
10 mL
Conduct applications accurately with Thermo Scientific™ Nalgene™ Polypropylene Graduated Cylinders, which feature generous pour spouts, molded-in graduations and a large blue base. There is no meniscus to confuse readings, eliminating guesswork; the cylinders can be chemically sterilized and are suitable for food and beverage use. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Excellent chemical resistance for a wide variety of solutions
  • Resists temperatures up to 135°C (275°F) for a variety of uses
  • Autoclavable (autoclaving will affect accuracy)
  • Molded-in graduations are silk-screened for easy reading (10mL to 1L sizes only)
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Ordering Information:
Individually packaged.
Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.
Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material.  Packaging bags are made from LDPE and are readily recyclable once the label is removed.   Please continue the cycle and recycle your cartons and bags.
.
Specifications
Capacity (Metric)1000 mL
MaterialPolypropylene
Tolerance±6 mL
Item DescriptionGraduated Cylinder PP 1000mL
Certifications/CompliancePart 177.1520, ASTM E1272, CFR 21, ISO 6706
No. per Pack1
No. per Case6
CalibrationTo Contain, To Deliver
Diameter (English)3.15 in.
Scale30mL to 1000mL in 10mL increments
Diameter (Metric)80 mm
IncludesBlue Polypropylene Base
Graduations10 mL
AutoclavableAutoclavable
Limit of Error±6.0mL
Max. Temperature121°C
ShapeRound
Diameter (Metric) Inside62 mm
Diameter (English) Inside2.44 in.
Max. Temperature (English)250°F
Base TypeHexagonal
Height (English)17.7 in.
Height (Metric)449.6 mm
TypeGraduated Cylinder
ACT LabelYes
Unit Size6