Benchtop Coolers, 4 x 5, -20C
Benchtop Coolers, 4 x 5, -20C
Actual product may vary
Benchtop Coolers, 4 x 5, -20C
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Catalog number 355501
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Height (Metric)133 mm
Height (English)8.25 in.
Length (Metric)226 mm
Length (English)8.94 in.
Width (Metric)165 mm
Width (English)6.5 in.
Dimensions (L x W x H)8.94 x 6.5 x 8.25 in. (226 x 165 x 133 mm)
Description4x5 compartments, -20 deg.
Array4 x 5
Holds Temp., Hours<-15°C, 1
DesignNon-filled/clear lid
MaterialPolycarbonate
TypeBenchtop Cooler
ColorClear
Capacity (Metric)0.2 to 0.5 mL
Holds20 (0.2 to 0.5mL Tubes)
Temperature Range-20°C
Unit Size1