Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Actual product may vary
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Thermo Fisher Scientific
Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentCertifiedVolume (Metric) WorkingBarcode
3455Immulon 2 HBNo280 μLNo Barcode
3855Immulon 4 HBXNo280 μLNo Barcode
475094PolySorpNo350 μLNo Barcode
Catalog number 3455
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Surface Treatment:
Immulon 2 HB
Certified:
No
Volume (Metric) Working:
280 μL
Barcode:
No Barcode
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Specifications
BarcodeNo Barcode
CertifiedNo
FormatStandard
Detection MethodColorimetric
Array8 x 12
Packaging50 Cases
Binding TypeSlightly Hydrophilic
ColorClear
Product TypeMicroplate
MaterialPolystyrene
Volume (Metric) Well330 μL
No. of Wells96
Volume (Metric) Working280 μL
SterilityNon-sterile
Surface TreatmentImmulon 2 HB
Well ShapeRound, Flat Bottom (F-well)
No. per Case50
No. per Pack10
Plate BlockingNone
Quantity50/Pk.
Target MoleculeProteins
Unit Size50