Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Actual product may vary
Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Thermo Fisher Scientific
Nalgene™ Square PET Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with these square bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
342040-0250250 mL
342040-0650650 mL
Catalog number 342040-0250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Provide excellent gas barrier properties for storage and shipping of liquid media, buffers and sera with Thermo Scientific™ Nalgene™ Square PET Media Bottles with Closure. These light-weight bottles are shatter-resistant, sterile and come in a space-saving square design.
Features:
  • Polyethylene terephthalate with natural high density polyethylene closure
  • Molded in graduations and neck ring for use with tamper evident shrink sleeves
  • Gamma irradiated to 10-6 Sterility Assurance Level (SAL)
  • Non-pyrogenic and non-cytotoxic
  • Transparency and reduced permeability to CO2 and O2
  • Bottle and closure material are Animal Derived Component (ADC) Free 
  • Leakproof†
Ordering Information:
Products are packaged in shrink-wrap trays. Trays are double polybagged inside the master carton.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)250 mL
Capacity (English)8.5 oz.
ColorClear
Diameter (English)2.3 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
MaterialPET
No. per Case60
No. per Pack30
SterilitySterile
Certifications/CompliancePET Media Bottles and HDPE closure materials meet current biological testing standards including USP Class VI, USP <661.1>
Closure IncludedYes
GraduatedYes
MouthNarrow
ShapeSquare
Fit for PurposeSterile
Height (Metric)140 mm
Product TypeMedia Bottle
Height (English)5.5 in.
Closure Size38-430 mm
Diameter (Metric)59 mm
Unit Size60