Trays for Precision™ Water Baths
Trays for Precision™ Water Baths
Actual product may vary
Trays for Precision™ Water Baths
Trays for Precision™ Water Baths
Thermo Fisher Scientific
Trays for Precision™ Water Baths
Hold test tubes and flasks in place with trays for Precision water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)
3161599Flask Tray 25mLPrecision Shaking Water Baths
3166228Flask Tray 50mLPrecision Shaking Water Baths
Catalog number 3161599
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Flask Tray 25mL
For Use With (Equipment):
Precision Shaking Water Baths
Hold test tubes and flasks in place with trays for Thermo Scientific™ Precision™ water baths.
Test Tube Tray
  • Measures 5 x 10.2in. (12.7 x 25.9cm)
  • Holds 10-13 25mm test tubes
Flask Trays
  • Measures 12.7 x 25.9cm (5 x 10.2in)
  • 25mL flask tray holds 18 clips
  • 50mL flask tray holds 10 clips
Specifications
DescriptionFlask Tray 25mL
Capacity (Metric)25 mL
For Use With (Equipment)Precision Shaking Water Baths
Shaking MotionReciprocating
TypeFlask Tray
Holds18 x 25 mL Flasks