Septa
Septa
Actual product may vary
Septa
Septa
Septa
Septa
Septa
Use low bleed, robust BTO Septa for demanding GC-MS applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDiameter (Metric) InnerDescriptionQuantity
31303228Low Bleed Septum12.7 mmBTO; 12.7 mm dia.; Max. Temp. 400°C50 (Glass Jar)
31303211Low Bleed Septum17 mmBTO; 17 mm50 (Glass Jar)
31303230Low Bleed Septum11.5 mmBTO; 11.5 mm dia.; Max. Temp. 400°C50 (Glass Jar)
31303240Low Bleed Septum9 mmBTO Septa; 9 mm; Max. Temp. 400°C50 (Glass Jar)
31303233-BPLow Coring Septum11 mmBTO; 11 mm dia.; Max. Temp. 400°C50 (Blister Pk.)
HYP31303233-BPLow Coring Septum11 mmBTO; 11 mm dia.; Max. Temp. 400°C50 (Blister Pk.)
313P3240-BPLow Coring Septum9 mmMarathon; 9 mm dia.; Max. Temp. 350°C50 (Blister Pk.)
Catalog number 31303228
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Low Bleed Septum
Diameter (Metric) Inner:
12.7 mm
Description:
BTO; 12.7 mm dia.; Max. Temp. 400°C
Quantity:
50 (Glass Jar)
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionBTO; 12.7 mm dia.; Max. Temp. 400°C
For Use With (Equipment)Trace and Focus GC Instruments
MaterialSilicone
TypeLow Bleed Septum
Diameter (Metric) Inner12.7 mm
Quantity50 (Glass Jar)
Unit Size