MaxQ™ Shaker Universal Platforms
MaxQ™ Shaker Universal Platforms
Actual product may vary
Thermo Fisher Scientific
MaxQ™ Shaker Universal Platforms
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Dimensions (L x W)
30115BIUniversal Platform without Clamps, 18 x 30 in.MaxQ 3000, 500030 x 18 in. (76 x 45.7 cm)
30128Dual Stacked Universal Platform without Clamps, 18 x 18 in.MaxQ™ 2000, MaxQ™ 6000 Shaker18 x 18 in. (45.7 x 45.7 cm)
Catalog number 30115BI
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Universal Platform without Clamps, 18 x 30 in.
For Use With (Equipment):
MaxQ 3000, 5000
Dimensions (L x W):
30 x 18 in. (76 x 45.7 cm)
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ Shaker Universal Platforms. With exceptional innovation, reliability and accuracy, these platforms are the optimal choice for a busy lab.
For use with MaxQ Shakers as indicated. Choose the platform that best suits your needs.
Ordering Information:
Purchase clamps separately.
Specifications
Product TypeShaker Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)30 x 18 in. (76 x 45.7 cm)
Length (English) Platform30 in.
Length (Metric) Platform76 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
DescriptionUniversal Platform without Clamps, 18 x 30 in.
For Use With (Equipment)MaxQ 3000, 5000
Unit Size