Nunc™ 96-Well Cap Mats
Nunc™ 96-Well Cap Mats
Actual product may vary
Nunc™ 96-Well Cap Mats
Nunc™ 96-Well Cap Mats
Nunc™ 96-Well Cap Mats
Nunc™ 96-Well Cap Mats
Thermo Fisher Scientific
Nunc™ 96-Well Cap Mats
Protect all 96 wells during short-term or long-term storage of chemicals or compounds with a single device using these well cap mats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityPlate CompatibilityNo. per Case
276002NaturalNon-sterileNunc 96 Round Well50
9503230NaturalNon-sterileNunc 96 Square Well50
Catalog number 276002
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Natural
Sterility:
Non-sterile
Plate Compatibility:
Nunc 96 Round Well
No. per Case:
50
Protect all 96 wells during short-term or long-term storage of chemicals or compounds with a single device using Thermo Scientific™ Nunc™ 96-Well Cap Mats, which prevent contamination and evaporation of well contents.
Features:
  • Chemically resistant and pierceable; can withstand low temperature to -80°C
  • A variety of colors for quick identification of storage plates
Pre-slit silicone well caps (276011 only):
  • Permit a pipette tip or probe to enter the well without damage to the cap
  • Solvent resistant, should not be subjected to 100% DMSO
Ordering Information:
Sterile mats are individually wrapped.
Note:
Designed specifically for use with microplates featuring 'shared wall' technology. 96-well polypropylene DeepWell™ plates with “shared wall” technology also fit standard-height polypropylene 96-well microwell plates. 276011 pre-slit silicone caps can be pierced repeatedly. Do not use pre-slit silicone with 100% DMSO.
Specifications
ColorNatural
Solvent Or Alcohol ResistantAlcohol Resistant, Solvent Resistant
MaterialTPE
SterilityNon-sterile
AutoclavableAutoclavable
No. of Wells96
No. per Case50
No. per Pack5
For Use With (Application)For compound storage to −80°C
Pre-slitNo
Product TypePlate Cover
Product LineNunc
AdhesiveNone
Certified RNase/DNase-freeYes
PierceableYes (Hypodermic)
Temperature Range (Metric)-20°C to +120°C
Well ShapeRound
Plate CompatibilityNunc 96 Round Well
Unit Size50