Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Sterility | Packaging Type | Quantity |
---|---|---|---|---|
263-1S | 10.7 mL | Sterile | Individually Wrapped | 100/Pk 8Pk/Cs 800/Cs |
FKS15357853 | 5.6 mL | Non-sterile | Bulk | 400/Pk 8Pk/Cs 4000/Cs |
FIS13-711-40 | 10.7 mL | Non-sterile | Bulk | 100/Pk 10Pk/Cs 1000/Cs |
FIS02-708-030 | 10.7 mL | Sterile | Individually Wrapped | 100/Pk 8Pk/Cs 800/Cs |
FKS15397853 | 10.7 mL | Sterile | Individually Wrapped | 100/Pk 8Pk/Cs 800/Cs |
FKS11651839 | 23 mL | Non-sterile | White Box | 100/Pk. |