Search Thermo Fisher Scientific
Catalog Number | Description | Dimensions (L x W) | For Use With (Equipment) |
---|---|---|---|
238000 | Universal Platform without Clamps, 18 x 18 in. | 18 x 18 in. | MaxQ™ HP Orbital Shakers |
238061 | Universal Platform without Clamps, 18 x 29.5 in. | 18 x 29.5 in. (74.9 x 45.7 cm) | MaxQ 8000 |
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms.