7961 Series Thermal Transfer Printer Ribbon
7961 Series Thermal Transfer Printer Ribbon
Actual product may vary
Thermo Fisher Scientific
7961 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent scratch and good chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)Diameter (Metric)
236013300 m60 mm25.4 mm
236014300 m110 mm25.4 mm
236015300 m114 mm25.4 mm
236016300 m170 mm25.4 mm
Catalog number 236013
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
300 m
Width (Metric):
60 mm
Diameter (Metric):
25.4 mm
When printed on the appropriate Brady™ label, this ribbon gives excellent scratch and good chemical resistance.
Specifications
ColorBlack
For Use With (Equipment)BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer
MaterialResin
Length (English)984 ft.
Width (English)2.4 in.
Length (Metric)300 m
Width (Metric)60 mm
Diameter (English)1 in.
Diameter (Metric)25.4 mm
FormatOpen-core Ribbon
Certifications/Compliance2011/65/EU and 2015/863
Alternate Part NumberR7961-60X300/O
Unit Size