Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Actual product may vary
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Perform precise dispensing of small drops into small-volume containers using these fine-tip transfer pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Bulb Draw (Metric)SterilityPackaging Type
232-1S5.8 mL3.3 mLSterileIndividually Wrapped 400/Pk 8Pk/Cs 3200/Cs
FKS116118391.5 mL1.0 mLNon-sterile
FIS13-711-251.7 mL1.0 mLNon-sterileBulk 400/Pk 8Pk/Cs 3200/Cs
FKS103748505.4 mL3.1 mLNon-sterileBulk 250/Pk 8Pk/Cs 2000/Cs
FIS13-711-265.8 mL3.3 mLNon-sterile500/Pk., 5000/Cs.
FIS13-711-315.8 mL3.7 mLNon-sterileBulk 500/Pk 10Pk/Cs 5000/Cs
Catalog number 232-1S
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5.8 mL
Bulb Draw (Metric):
3.3 mL
Sterility:
Sterile
Packaging Type:
Individually Wrapped 400/Pk 8Pk/Cs 3200/Cs
These fine-tip transfer pipettes are ideal for removing separating liquid samples. Designed with a built-in pipette bulb, they allow precise dispensing of small drops into small-volume containers.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface – prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Available with fine tip or extended fine tip
Specifications
Bulb Draw (Metric)3.3 mL
DescriptionFine Tip Standard Bulb Transfer Pipette, 5.8mL
Diameter (Metric) Outer4.6 mm
MaterialLDPE
SterilitySterile
Drops/mL53
GraduatedNo
Packaging TypeIndividually Wrapped 400/Pk 8Pk/Cs 3200/Cs
Volume (Metric)5.8 mL
Length (Metric)14.7 cm
TypeFine Tip Transfer Pipet