Reverse Osmosis Membranes
Reverse Osmosis Membranes
Actual product may vary
Thermo Fisher Scientific
Reverse Osmosis Membranes
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
22.0046Reverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
22.0087Reverse Osmosis membrane
09.2003Reverse Osmosis membrane with integrated pretreatment, 3L/hr
09.2006Reverse Osmosis membrane with integrated pretreatment, 6L/hr
Catalog number 22.0046
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Reverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
  • Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
  • Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
  • LabTower™ EDI 15 requires two of these RO membranes
  • LabTower™ RO 20 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • LabTower™ TII 20 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
  • Pacific™ RO 3, 7, 12 require one of these RO membranes
  • Pacific RO 20 requires two of these RO membranes
  • Pacific™ TII 3, 7, 12 require one of these RO membranes
  • Pacific TII 20 requires two of these RO membranes
22.0087:
  • LabTower™ EDI 30 requires two of these RO membranes
  • LabTower™ RO 40 requires two of these RO membranes
  • LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • LabTower TII 40 requires two of these RO membranes
  • LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
  • Pacific RO 40 requires two of these RO membranes
  • Pacific TII 40 requires two of these RO membranes
Specifications
DescriptionReverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
For Use With (Equipment)LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20
TypeReverse Osmosis Membrane
Unit Size