SafeBasics™ Procedure Earloop Face Masks
SafeBasics™ Procedure Earloop Face Masks
Actual product may vary
Thermo Fisher Scientific
SafeBasics™ Procedure Earloop Face Masks
Ultrasonically sealed procedure face masks are soft and stretchy with latex-free earloops and an adjustable nosepiece to ensure a comfortable fit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberdimColordimASTMLevel
2130
2140
Catalog number 2130
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Primary protection for simple, low-risk procedures involving very light splatter, moisture, spray and spatter. Ideal for patient appointments and exams
  • Inner and outer layer constructed for basic fluid resistance, with ultrasonically sealed, soft, stretchy, latex free earloops
  • Adjustable nosepiece for custom and comfortable fit and easy breathing
  • Economic alternative for simple procedures
  • ASTM Level 1, 2 and 3 protection
Specifications
Packaging Quantity50 Masks/Box, 10 Boxes/Cs.
Unit Size