Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Actual product may vary
Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Thermo Fisher Scientific
Nalgene™ Square Wide-Mouth HDPE Bottles with Closure
Easily fill these square, wide-mouth HDPE bottles that offer excellent chemical resistance, making them suitable for a wide variety of uses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2114-0016500 mL53 mm
2114-00321 L63 mm
Catalog number 2114-0016
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Closure Size:
53 mm
Easily fill Thermo Scientific™ Nalgene™ Square Wide-Mouth HDPE Bottles. HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses. Good for freezer storage to -100°C (-148°F). This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
  • Wide mouth is easy to fill
  • Bottle design saves space in freezer
  • HDPE exhibits excellent chemical resistance to most acids, bases and alcohols, making these bottles suitable for a wide variety of uses
  • Linerless caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
DescriptionWide-Mouth Square Bottle
Diameter (English)2.9 in.
Diameter (English) Inner Neck1.7 in.
Diameter (Metric) Inner Neck43 mm
MaterialHDPE
No. per Case48
No. per Pack12
SterilityNon-sterile
Closure ColorNatural, translucent
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeSquare
ACT LabelYes
Height (Metric)146 mm
Product TypeSquare Wide Mouth Bottle
Width (English)2.4 in.
Height (English)5.7 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size53 mm
Diameter (Metric)74 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit Size48