Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Actual product may vary
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Nalgene™ Narrow-Mouth HDPE Economy Bottles, 500mL
Contain samples and daily working solutions like buffers and bench reagents with these HDPE economy bottles, suitable for lighter-duty, general purpose applications.
Have Questions?
Catalog number 2089-0016
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Contain samples and daily working solutions like buffers and bench reagents with Thermo Scientific™ Nalgene™ Narrow-Mouth HDPE Economy Bottles, lightweight versions of the original Nalgene Lab Quality Bottle. Suitable for lighter-duty, general purpose applications, these high-density polyethylene bottles are guaranteed leakproof†. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
  • Linerless polypropylene caps ensure leakproof† performance without a liner that can wrinkle, cause leaks and contaminate your solutions
  • Recommended for sample collection and storage of daily working solutions
  • Bottles participate in My Green Labs' ACT Label program for Accountability, Consistency and Transparency around product sustainability
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size28 mm
Closure TypeScrew Cap
Diameter (Metric)73 mm
Height (English)2.4 in.
Height (Metric)170 mm
Through the Closure DispensingNo
Product TypeNarrow Mouth Economy Bottle
Capacity16 oz., 500 mL
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorTranslucent
DescriptionNarrow-Mouth Economy Bottle
Diameter (Metric) Inner Neck21 mm
For Use With (Application)Lighter-Duty, General Purpose
MaterialHDPE
No. per Case48
No. per Pack48
SterilityNon-sterile
Closure ColorNatural
Neck Finish28 mm
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
MouthNarrow
ShapeRound
ACT LabelYes
Unit Size48