ART™ Barrier Extended Length Pipette Tips
ART™ Barrier Extended Length Pipette Tips
Actual product may vary
ART™ Barrier Extended Length Pipette Tips
ART™ Barrier Extended Length Pipette Tips
Thermo Fisher Scientific
ART™ Barrier Extended Length Pipette Tips
Perform pipetting accurately and consistently with Thermo Scientific™ ART™ Extended Length Barrier Pipette Tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelTip StyleFit for Purpose
2079-HR1000 μLART BarrierUniversal FitSterile and PCR Ready
2140-HR10 μLART 10 REACHExtended LengthSterile and PCR Ready
2140-05-HR10 μLART 10 REACHLow RetentionSterile and PCR Ready
2160P-HR200 μLART BarrierUniversal FitSterile and PCR Ready
2079-05-HR1000 μLART BarrierLow RetentionSterile and PCR Ready
2179-05-HR1000 μLART 1000LLow RetentionSterile and PCR Ready
Catalog number 2079-HR
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
1000 μL
Model:
ART Barrier
Tip Style:
Universal Fit
Fit for Purpose:
Sterile and PCR Ready
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks and the covers may be used as hinged or lift-off style. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. ART hinged racks are available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Stackable hinged racks save space in your lab
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
DescriptionART Barrier, Filtered, Sterile, Hinged Rack
Volume (Metric)1000 μL
FilteredFiltered
SterilitySterile
ColorNatural
ModelART Barrier
Length (English)4.7 in.
Length (Metric)11.85 cm
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
FormatHinged Rack
Tip StyleUniversal Fit
For Use With (Equipment)Universal Fit
Product LineART
Fit for PurposeSterile and PCR Ready
Unit Size768