Samco™ General-Purpose Transfer Pipettes, 7.7 mL
Product Image
Samco™ General-Purpose Transfer Pipettes, 7.7 mL
Prevent cross-contamination with these transfer pipettes, which have a built-in bulb that eliminates the need for plugging.
Have Questions?
Catalog number 202-1S
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Prevent cross-contamination with Thermo Scientific™ Samco™ Transfer Pipettes, which have a built-in bulb that eliminates the need for plugging, packaging and autoclaving. Samco transfer pipettes are made of shatterproof, nontoxic polyethylene. Their low-affinity surface prevents proteins binding in research applications. They can be frozen in liquid nitrogen, or heat sealed for storage.
Recommended for: Research Applications
Specifications
Bulb Draw (Metric)3.5 mL
DescriptionGeneral Purpose Transfer Pipettes, Standard Bulb, 7.7mL
Diameter (Metric) Outer7.9 mm
MaterialLDPE
SterilitySterile
Drops/mL21
GraduatedNo
Packaging TypeIndividually Wrapped 500/Pk
Volume (Metric)7.7 mL
Length (Metric)15.6 cm
TypeGeneral Purpose Transfer Pipet
DisposableSingle-use
Unit Size4000