Cryo Aprons
Cryo Aprons
Actual product may vary
Cryo Aprons
Cryo Aprons
Thermo Fisher Scientific
Cryo Aprons
Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
189452Medium
189453Large
Catalog number 189452
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Medium
Protect your clothing with Thermo Scientific™ Cryo Aprons, available in small, medium and large sizes.
  • Small: 36 in. (91.4cm)
  • Medium: 42 in. (106.7cm)
  • Large: 48 in. (121.9cm)
  • Warranty varies by country, please contact us for details
Specifications
TypeCryogenic Apron
SizeMedium
ColorBlue
Width (English)42 in.
Width (Metric)106.7 cm
Unit Size